Learn More
Thermo Scientific™ Nalgene™ Square PETG Media Bottles with Closure: Nonsterile, Shrink-Wrapped Trays

Description
Package and ship liquid media, buffers and sera using Thermo Scientific™ Nalgene™ PETG Square Media Bottles with Closures. Heavy-walled, durable bottles offer excellent gas barrier properties and are specifically designed for storing media and sera.
- Heavy-walled and durable
- Shatter-resistant
- Provide excellent gas barrier properties
- In shrink-wrap trays
- Molded in graduations
- Clear bottles for easy identification of contents
- Nonsterile
- Leakproof†
Note:
† The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
- Environmental benefits include:
- Renewable Energy

Specifications
Specifications
Capacity (Metric) | 250 mL |
Capacity (English) | 8.5 oz. |
Color | Clear |
Description | 250mL, 38-430 closure, tray pack |
Diameter (English) | 2.4 in. |
Diameter (English) Inner Neck | 1.1 in. |
Diameter (Metric) Inner Neck | 28 mm |
For Use With (Application) | Storing Media and Sera |
Material | PETG |
No. per Case | 60 |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
1422896 | Certificate of Compliance | 12/02/2025 | 322020-0250 |
1419522 | Certificate of Compliance | 27/12/2024 | 322020-0250 |
1304744 | Certificate of Compliance | 10/12/2024 | 322020-0250 |
1300847 | Certificate of Compliance | 10/12/2024 | 322020-0250 |
1400079 | Certificate of Compliance | 26/01/2024 | 322020-0250 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.