Learn More
Thermo Scientific™ Nalgene™ Square PETG Media Bottles with Closure
Description
Eliminate costly washing, depyrogenation and autoclaving steps with certified sterile and non-pyrogenic Thermo Scientific™ Nalgene™ Square PETG Storage Bottles with Closure. Ideal for storage and sampling of active pharmaceutical ingredients and bulk intermediates, and also well suited to the preparation and containment of buffers, cultures or prolonged storage of pH-sensitive liquids such as culture media.
Features:
- Made from durable, break-resistant polyethylene terephthalate copolyester, glycol modified (PETG)
- Suitable for frozen storage down to -40°C
- Heat-shrink band around closure and neck for tamper-evident seal
- Leakproof bottles and HDPE closures provided
- Sterile and non-pyrogenic
- Available in low particulate format
- Supported by extensive validation documentation
Ordering Information:
- Packed in shrink-wrapped trays
Note:
- 2 L size has molded-in handgrips
Warranty:
- 90 days

Specifications
Specifications
Capacity (Metric) | 2000 mL |
Capacity (English) | 64 oz. |
Color | Clear |
Diameter (English) | 4.57 in. |
Diameter (English) Inner Neck | 1.5 in. |
Diameter (Metric) Inner Neck | 39 mm |
For Use With (Application) | Media/Buffer Preparation , Media/Buffer Storage, Storage |
Material | PETG |
No. per Case | 12/Cs. |
No. per Pack | 6/Pk. |
Show More |
Documents
Product Information
Schematics & Diagrams
Scientific Resources
Application Notes
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
1424435 | Certificate of Processing | 25/04/2025 | 2019-2000 |
1424140 | Certificate of Processing | 21/04/2025 | 2019-2000 |
1424139 | Certificate of Processing | 01/04/2025 | 2019-2000 |
1426955 | Certificate of Compliance | 17/04/2025 | 2019-2000 |
1424435 | Certificate of Compliance | 09/04/2025 | 2019-2000 |
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.