missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Separatory Funnels made with Teflon™ fluoropolymer with Closure made with Tefzel™
GREENER_CHOICE

Utilize these chemically inert, translucent and autoclavable funnels which feature a TFE stopcock assembly. Chemical-resistant, leakproof and requires no lubrication.

Brand:  Thermo Scientific™ 4301-2000

864.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 10120551

  • 899.00 € / Pack of 2
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Perform your liquid-liquid extractions with these chemically inert, translucent and autoclavable Thermo Scientific™ Nalgene™ Separatory Funnels made with Teflon™ fluoropolymer and Closure made with Tefzel™ which feature a TFE stopcock assembly. TFE stopcock assembly has excellent chemical resistance, is leakproof† when used with provided Nalgene closure and does not require lubrication — can be chemically disinfected. More transparent than our polypropylene separatory funnels. Compatible with all common lab reagents and solvents. Won't chip, crack or shatter like glass separatory funnels.

Features:
  • Break-resistant and impervious to any chemical used in a separatory funnel
  • No phthalates to contaminate analyses
  • Useful in radiological and hazardous separations
  • Autoclavable/Leakproof
  • Stem length: 65mm

Includes:

Teflon™ FEP funnel, TFE stopcock and Tefzel™ ETFE screw closure

Ordering Information:

For replacement stopcock assembly for 125mL to 1L sizes, order Cat. No. 714301-1025. For replacement stopcock assembly for 2L size, order Cat. No. 714301-1050.

Note:

Funnel can be autoclaved; stopcock can be chemically sterilized. No stopcock lubrication needed.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

2000 mL
401 mm
1/Pk.; 2/Cs.
Teflon FEP
Separatory Funnel FEP 2000mL; 53mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number1425385 Certificate TypeCertificate of Compliance Date17/04/2025 Catalog Number4301-2000
Lot Number1420289 Certificate TypeCertificate of Compliance Date08/02/2025 Catalog Number4301-2000
Lot Number1317106 Certificate TypeCertificate of Compliance Date09/12/2024 Catalog Number4301-2000
Lot Number1404925 Certificate TypeCertificate of Compliance Date17/04/2024 Catalog Number4301-2000
Lot Number1390720 Certificate TypeCertificate of Compliance Date09/09/2023 Catalog Number4301-2000
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Nalgene™ Separatory Funnels made with Teflon™ fluoropolymer with Closure made with Tefzel™ > 2000mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

Teflon™ is a trademark of The Chemours Company FC, LLC used under license by Nalge Nunc International Corporation.

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2 psig for 2 minutes, and no water escapes. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes.

Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.