Learn More
Thermo Scientific™ Nalgene™ Torque Wrench Fittings for ETFE Closures
Assure accurate torque readings when applying closures onto bottles with Nalgene™ Torque Wrench Fittings for ETFE closures.
129.00 € - 298.00 €
Specifications
Product Type Fittings
Drive Size 1/4 in.
For Use With (Equipment) Nalgene Torque Wrenches (Cat. No. 2195-6169)
Material Epoxy
No. per Pack 1 Ea.
View More Specs
Products 9
Product Code Brand Closure Size Price Quantity & Availability  
Product Code Brand Closure Size Price Quantity & Availability  
11994932
View Documents
Thermo Scientific™
2195-2020
20 to 415 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11904942
View Documents
Thermo Scientific™
2195-2024
24 to 415 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11914942
View Documents
Thermo Scientific™
2195-2028
28 to 415 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11924942
View Documents
Thermo Scientific™
2195-2033
33 to 415 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11934942
View Documents
Thermo Scientific™
2195-2038
38 to 415 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11944942
View Documents
Thermo Scientific™
2195-2043
43 to 415 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11954942
View Documents
Thermo Scientific™
2195-2048
48 to 415 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11964942
View Documents
Thermo Scientific™
2195-2053
53 to 415 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11974942
View Documents
Thermo Scientific™
2195-2438
38 to 430 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
Assure accurate torque readings when applying closures onto bottles with Thermo Scientific™ Nalgene™ Torque Wrench Fittings for ETFE Closures. Proper application torque is essential for leakproof† performance.
  • Durable epoxy resin construction
  • Specifically designed for Nalgene ETFE closures
Compatible with:
Compatible with Thermo Scientific Nalgene torque wrench 2195-6169 † The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
Warranty: 90 days
Specifications
Specifications
Fittings
Nalgene Torque Wrenches (Cat. No. 2195-6169)
1 Ea.
1/4 in.
Epoxy
Videos
SDS
Documents
Documents
Product Certifications
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.