missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific™ Nalgene™ Unitary™ LDPE Wash Bottles Product Code.: 10558701
Add to basket

Thermo Scientific™ Nalgene™ Unitary™ LDPE Wash Bottles
GREENER_CHOICE

Product Code. 10558701
125 mL, 24 mm
Click to view available options
Capacity (Metric):
125 mL
250 mL
500 mL
750 mL
1000 mL
Closure Size:
24 mm
38 mm
43 mm

Product Code. 10558701

Brand: Thermo Scientific™ 24020125

Edge
Add to basket
Add to basket

Dispense contents without tipping or shaking by using these LDPE wash bottles, designed with a wide mouth for faster, easier filling.

Dispense contents without tipping or shaking with Thermo Scientific™ Nalgene™ Unitary™ LDPE Wash Bottles. Wide mouth allows faster, easier filling. Bottle and tubulation are molded in one piece for uniform service and precision flow without sputtering. The tip can be cut back to increase flow. Made from high-quality, laboratory-grade, plastic materials for dependably low leachables and extractables. Naturally flexible LDPE material is chemically compatible with a wide range of laboratory reagents and provides an ergonomic soft squeeze to help reduce hand fatigue.

Features:
  • Bottle and tubulation are molded in one piece for uniform leakproof† service
  • Tip delivers a uniform, ultrafine stream without sputtering, but can also be cut back to increase flow
  • Leakproof†

Includes:

Polypropylene screw closure

Greener Choice Claims
  • Environmental benefits include:
    • Renewable Energy
TRUSTED_SUSTAINABILITY

Specifications

Capacity 4 oz., 125 mL
Capacity (Metric) 125 mL
Capacity (English) 4 oz.
Color Translucent
Description LDPE bottle/tubulation
Material LDPE
No. per Case 48
No. per Pack 6
Closure Included Yes
Graduated No
Mouth Wide
Shape Cylindrical
Height (Metric) 119.4 mm
Product Type Wash Bottle
Height (English) 4.7 in.
Closure Material Polypropylene
Closure Size 24 mm
Diameter (Metric) 48.3 mm
Closure Type Screw Cap
Through the Closure Dispensing No
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number1285108 Certificate TypeCertificate of Compliance Date10/12/2024 Catalog Number2402-0125
Lot Number1303715 Certificate TypeCertificate of Compliance Date10/12/2024 Catalog Number2402-0125
Lot Number1413084 Certificate TypeCertificate of Compliance Date29/08/2024 Catalog Number2402-0125
Lot Number1392909 Certificate TypeCertificate of Compliance Date26/12/2023 Catalog Number2402-0125
Lot Number1398961 Certificate TypeCertificate of Compliance Date20/12/2023 Catalog Number2402-0125
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Nalgene™ Unitary™ LDPE Wash Bottles >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.