Learn More
Thermo Scientific™ National Vial Crushers
Reduce waste disposal costs by reducing the total volume of waste with the Thermo Scientific™ National Vial Crusher.
Brand:  Thermo Scientific™ C4040-VC
 View more versions of this product
Product Code. 10520136
  • 12892.36 € / Each
Estimated Shipment: 25-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Reduce waste disposal costs by reducing the total volume of waste with the Thermo Scientific™ National Vial Crusher.
  • Breaks and separates liquid from all 1mL to 100mL vials and ampules
  • Capable of processing approximately 5000 2mL vials per hour
  • Safe use in a fume hood or connect to a vacuum for limited exposure to flammable or toxic vapors
  • Environmentally responsible waste handling—crushed glass is automatically separated from solvent for separate disposal
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Manual Benchtop Vial Crusher, 241/2H x 81/2W x 231/2 in. D
National