Learn More
Thermo Scientific™ National Vial Crushers
Reduce waste disposal costs by reducing the total volume of waste with the Thermo Scientific™ National Vial Crusher.
12892.36 € - 16651.50 €
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
10129094
|
Thermo Scientific™
C4040-PVCM |
Pneumatic Benchtop Vial Crusher with Stainless-steel Mobile Cart | N/A | ||||||
10520136
|
Thermo Scientific™
C4040-VC |
Manual Benchtop Vial Crusher, 241/2H x 81/2W x 231/2 in. D |
12892.36 €
Each |
Estimated Shipment: 24-02-2025
Log in to see stock. |
|||||
Description
Reduce waste disposal costs by reducing the total volume of waste with the Thermo Scientific™ National Vial Crusher.
- Breaks and separates liquid from all 1mL to 100mL vials and ampules
- Capable of processing approximately 5000 2mL vials per hour
- Safe use in a fume hood or connect to a vacuum for limited exposure to flammable or toxic vapors
- Environmentally responsible waste handling—crushed glass is automatically separated from solvent for separate disposal
Specifications
National |