Learn More
Memmert™ Natural Convection Hot Air Sterilizer with SingleDISPLAY
Sterilize a variety of containers and instruments with this hot air sterilizer. Memmert™ Natural Convection Hot Air Sterilizers with SingleDISPLAY allow adjustment of pre-heated fresh air admixture by air flap control in 10% steps. Safer than disinfection.
Brand:  Memmert™ SN75
1491.75 EUR valid until 2024-12-15
Use promo code "21658" to get your promotional price.
 View more versions of this product
Product Code. 12885843
  • 1865.00 € / Each
Estimated Shipment: 19-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • German, English, Spanish, French, Polish, Czech, Hungarian language settings
  • SingleDISPLAY provides adaptive multifunctional digital PID-microprocessor controller with high-definition TFT-color display
  • Digital backwards counter with target time setting, adjustable from 1 minute to 99 days
  • Process time does not start until the set temperature is reached
  • Three freely selectable temperature values
  • Adjust temperature (Celsius or Fahrenheit), air flap position, program time, time zones, summertime/wintertime
Specifications
Specifications
CE, cUL, DVE, EAC
Natural Convection Hot Air Sterilizer
Applied for sterilisation of medical materials, complete killing of highly resistant microorganisms, special application of depyrogenisation
Stainless Steel
585 x 944 x 514 mm
585 mm
944 mm
514 mm
66 kg
115/230 V
50/60 Hz
74 L
TFT
Works Calibration Certificate
SN75
514 mm
400 mm
560 mm
330 mm
115/230 V 50/60 Hz
1800/2500 W
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products