missing translation for 'onlineSavingsMsg'
Learn More

RL Enterprises™ Natural Gas Security Bunsen Burner

Provides a single, continuous flame by controlled mixing of gas with air. RL Enterprises™ Natural Gas Security Bunsen Burner with NF Screwing G 1/2 Connector is manually adjustable to control intensity, temperature and flame size.

Brand:  RL Enterprises™ 4006/2TCM

Product Code. 11759777

  • 84.95 â‚¬ / Each
Only null left
Estimated Shipment: 14-10-2025
Add to basket

Description

Description

  • Large, stable base
  • Durable construction
  • Integral air regulator
Specifications

Specifications

Safety Bunsen Burner
13 mm
165 mm
1 Ea.
Natural Gas, Methane
Tétine Connector
T-Tine Connector
0.37 kg
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
RL Enterprises™ Natural Gas Security Bunsen Burner > Height: 165mm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.