missing translation for 'onlineSavingsMsg'
Learn More

Saint Gobain Life Sciences Natural Rubber Folding Skirt Stoppers

Use these stoppers for extra protection of tube ends during transport, handling and other operations during which tubes require both internal and end protection. Saint Gobain Life Sciences™ Natural Rubber Folding Skirt Stoppers provide an excellent seal.

Brand:  Saint Gobain Life Sciences 407020-20

 View more versions of this product

Product Code. 11714598

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Stoppers are flexible and provide a complete seal and good adhesion
  • Grey-blue grade: made from natural rubber
  • Temperature resistance from -35 to +70°C (120°C in moist heat)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

17.8 to 22.1mm
20/Pk.
Folding Skirt Stopper
12.5mmH/19mmH
Natural Rubber
19.4D
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.