Learn More
Saint Gobain Life Sciences Natural Rubber Folding Skirt Stoppers
Use these stoppers for extra protection of tube ends during transport, handling and other operations during which tubes require both internal and end protection. Saint Gobain Life Sciences™ Natural Rubber Folding Skirt Stoppers provide an excellent seal.
22.40 € - 45.10 €
Specifications
Material Natural Rubber
View More Specs
Products 6
Product Code Brand Height (Metric) Price Quantity & Availability  
Product Code Brand Height (Metric) Price Quantity & Availability  
11704598
View Documents
Saint Gobain Life Sciences
407018-20
11.5mmH/20mmH This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11784588
View Documents
Saint Gobain Life Sciences
407013-50
10mmH/20mmH This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11794588
View Documents
Saint Gobain Life Sciences
407015-20
11mmH/20mm H This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11714598
View Documents
Saint Gobain Life Sciences
407020-20
12.5mmH/19mmH This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11724598
View Documents
Saint Gobain Life Sciences
40702510
18mmH/25mmH This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11774588
View Documents
Saint Gobain Life Sciences
407010-50
8.5mmH/13mmH This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
  • Stoppers are flexible and provide a complete seal and good adhesion
  • Grey-blue grade: made from natural rubber
  • Temperature resistance from -35 to +70°C (120°C in moist heat)
Specifications
Specifications
Natural Rubber