missing translation for 'onlineSavingsMsg'
Learn More

Ansell Edmont™ Profil™ Plus 87-850 Series Yellow Natural Rubber Latex Immersion Gloves

Medium weight gloves with outstanding grip. Excellent resistance to mechanical hazards and enhanced chemical resistance.100% cotton flocklining makes the glove softer and more comfortable to wear while helping to absorb perspiration. Specially treated to reduce the risk of allergy.

34.55 €

Specifications

Coating Material Natural Rubber Latex
Thickness (Metric) 0.55 mm
Glove Cuff Style Pinked
View More Specs

Products 4
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
12435120
View Documents
Ansell Edmont™
87850080
8 N/A N/A N/A
12445120
View Documents
Ansell Edmont™
87850090
9 N/A N/A N/A
12425120
View Documents
Ansell Edmont™
87850070
N/A N/A N/A
12455120
View Documents
Ansell Edmont™
87850100
N/A N/A N/A
Description

Description

  • Specially treated to reduce the risk of allergy
  • Excellent resistance to mechanical hazards and enhanced chemical resistance
  • 100% cotton flocklining - soft, comfortable and absorbent
  • Ideal for janitorial use and handling in wet conditions
Specifications

Specifications

Natural Rubber Latex
0.55 mm
Pinked
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ansell Edmont™ Profil™ Plus 87-850 Series Yellow Natural Rubber Latex Immersion Gloves

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.