Learn More
Deutsch Neumann™ The Flip™ Safety Pipette Fillers
Reduces the risks of pipetting when handling toxic, corrosive, bacteria contaminated or bad smelling liquids. Deutsch Neumann™ The Flip™ Safety Pipette Fillers are manufactured from natural rubber.
Brand:  Deutsch Neumann™ 4710021
 View more versions of this product
Product Code. 10436511
  • 4.85 € / Each
Estimated Shipment: 26-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Only two operating points
  • Evacuate via the new automatic valve
  • Available in a range of colors
  • One-handed design is the simplest safety pipette filler available
  • Ability to clean inside of bulb by removing patented valve and rinsing out
Specifications
Specifications
Green
Natural Rubber
'Flip' Safety Pipette Filler
1 Each