Learn More
Deutsch Neumann™ The Flip™ Safety Pipette Fillers
Reduces the risks of pipetting when handling toxic, corrosive, bacteria contaminated or bad smelling liquids. Deutsch Neumann™ The Flip™ Safety Pipette Fillers are manufactured from natural rubber.
4.80 € - 4.85 €
Specifications
Material Natural Rubber
View More Specs
Products 2
Product Code Brand Color Price Quantity & Availability  
Product Code Brand Color Price Quantity & Availability  
10436511
View Documents
Deutsch Neumann™
4710021
Green
4.85 €
Each
Estimated Shipment: 30-12-2024
to see stock.
Add to basket
10473194
View Documents
Deutsch Neumann™
4710001
Red
4.80 €
Each
Estimated Shipment: 16-12-2024
to see stock.
Add to basket
Description
Description
  • Only two operating points
  • Evacuate via the new automatic valve
  • Available in a range of colors
  • One-handed design is the simplest safety pipette filler available
  • Ability to clean inside of bulb by removing patented valve and rinsing out
Specifications
Specifications
Natural Rubber