missing translation for 'onlineSavingsMsg'
Learn More

CleanSpace™ Technology CleanSpace™ Neck Pad

Thick foam neck pads are designed to be used with CleanSpace2 Respirators and can be attached to the unit using velcro strips.

Brand:  CleanSpace™ Technology PAF-0014

 View more versions of this product

Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 15930052

  • 80.05 € / Each
Estimated Shipment: 23-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Used with the CleanSpace2 Respirators: light weight, no hoses or belts
  • Designed for comfort over long periods
  • Allows sweating and offers breathability
  • Easy to wash and quick drying; quick to replace
  • Designed for long wear in harsh environments
  • Hygienic (medical quality materials) and hypoallergenic
Specifications

Specifications

Reusable
Comfort Pad
Welding, Woodworking, Manufacturing, Smelting, Construction, Mining, Agriculture, Grinding
PAF-0014
163 x 95 x 30 mm
Foam
Thick
CleanSpace 2 Respirators
Store at -10°C to +55°C
20 g
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers