Learn More
Tocris Bioscience™ Ned K
NAADP signaling inhibitor
Brand:  Tocris Bioscience™ 5976/50
1180.96 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
 View more versions of this product
Product Code. 18764039
  • 1330.00 € / 50mg
Explore more special offers
Description
Description
Ned K is a NAADP inhibitor. Prevents Ca2+ oscillations in rat cardiomyocytes in vitro during simulated ischemia and reperfusion, but exhibits no significant effect on Ca2+ sparks. Significantly reduces infarct size in rat cardiac ischemia/reperfusion model in vivo.
Specifications
Specifications
Cream
C31H31N5O3
Soluble to 50mM in DMSO
Prevents Ca2+ oscillations
Inhibitor
521.61g/mol
Store at -20°C