missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Ned K

NAADP signaling inhibitor

331.00 € - 1330.00 €

Specifications

Color Cream
Product Type Inhibitor
Molecular Formula C31H31N5O3
Formula Weight 521.61g/mol
Solubility Soluble to 50mM in DMSO
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18776828
View Documents
Tocris Bioscience™
5976/10
10 mg
331.00 €
10mg
Please to purchase this item. Need a web account? Register with us today!
18764039
View Documents
Tocris Bioscience™
5976/50
50 mg
1330.00 €
50mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Ned K is a NAADP inhibitor. Prevents Ca2+ oscillations in rat cardiomyocytes in vitro during simulated ischemia and reperfusion, but exhibits no significant effect on Ca2+ sparks. Significantly reduces infarct size in rat cardiac ischemia/reperfusion model in vivo.
Specifications

Specifications

Cream
C31H31N5O3
Soluble to 50mM in DMSO
Prevents Ca2+ oscillations
Inhibitor
521.61g/mol
Store at -20°C
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only