Learn More
Dr.Weigert™ Neodisher LaboClean FLA
Neodisher LaboClean FLA - Alkaline detergent, liquid concentrate for the automated cleaning of commonly used laboratory utensils (in medical, biological, microbiological, chemical, food, pharmaceutical, ... ). Removes stubborn oily residues and other organic residues, e.g. fats, oils, starch, residues of felt-tip pens
Brand:  Dr.Weigert™ 411230
 View more versions of this product
Product Code. 11758373
  • 128.00 € / 10 L
Explore more special offers
Description
Description
  • Alkaline detergent, liquid concentrate
  • neodisher LaboClean FLA is used for cleaning in washer disinfection
  • Free of phosphates, surfactants and active chlorine
  • Removes organic residues, especially suitable for cleaning animal cages for mice or rats made of polysulfone, polycarbonate, polyetherimide, polyphenylsulfone or stainless steel
  • Dosage of between 2-6 mL/L
Specifications
Specifications
Neodisher FLA
12.3 to 12.8
Polycarboxylates, potassium hydroxide
Canister
310 x 230 x 180 mm
10 L
Liquid
Non-sterile
1 to 6 mL/L
1.4 g/cm3 (20°C)
Removes Stubborn Oily Residues and Other Organic Residues
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Supplemental information
  • MIXTURE LIST-Contains : potassium hydroxide