missing translation for 'onlineSavingsMsg'
Learn More

Dr.Weigert™ Neodisher LaboClean FLA

Neodisher LaboClean FLA - Alkaline detergent, liquid concentrate for the automated cleaning of commonly used laboratory utensils (in medical, biological, microbiological, chemical, food, pharmaceutical, ... ). Removes stubborn oily residues and other organic residues, e.g. fats, oils, starch, residues of felt-tip pens

77.05 € - 128.00 €

Specifications

Product Type Neodisher FLA
Form Liquid
pH 12.3 to 12.8
Sterility Non-sterile
Container Type Canister
View More Specs

Products 2
Product Code Brand Volume (Metric) Price Quantity & Availability  
Product Code Brand Volume (Metric) Price Quantity & Availability  
11758373
View Documents
Dr.Weigert™
411230
10 L
128.00 €
10 L
Please to purchase this item. Need a web account? Register with us today!
11768373
View Documents
Dr.Weigert™
411233
5 L
77.05 €
5 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Alkaline detergent, liquid concentrate
  • neodisher LaboClean FLA is used for cleaning in washer disinfection
  • Free of phosphates, surfactants and active chlorine
  • Removes organic residues, especially suitable for cleaning animal cages for mice or rats made of polysulfone, polycarbonate, polyetherimide, polyphenylsulfone or stainless steel
  • Dosage of between 2-6 mL/L
Specifications

Specifications

Neodisher FLA
12.3 to 12.8
Canister
Liquid
Non-sterile
Removes Stubborn Oily Residues and Other Organic Residues
Videos
Safety and Handling

Safety and Handling

Supplemental information
  • MIXTURE LIST-Contains : potassium hydroxide

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.