Learn More
Neodymium Boron Iron, REacton™, Thermo Scientific Chemicals
Brand:  Thermo Scientific Alfa Aesar 013642.30
 View more versions of this product
Product Code. 11316898
  • 383.91 € / 250g
Explore more special offers
Description
Description
Neodymium Boron Iron applied to manufacture head actuators for computer hard disks, magnetic resonance imaging (MRI), magnetic guitar pickups, loudspeakers and headphones, mechanical e-cigarette firing switches and magnetic bearings and couplings.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Neodymium Boron Iron
Gray-Silver
250 g
MFCD02091729
Insoluble in water.
Silver gray broken lump
Magnet Alloy Additive
Broken Lump
Nd:B:Fe; 15:8:77 atomic%
Magnet Alloy Additive
Odorless