missing translation for 'onlineSavingsMsg'
Learn More

Ansell™ NeoTouch™ 25-101 Series Bright Green Neoprene Gloves

With a latex-free formulation, NeoTouch™ gloves are appropriate for the prevention of Type I allergies. Because they’re powder-free, they also limit the risk of dermatitis for the wearer. They provide excellent resistance to acids, bases and alcohols.

29.50 € - 30.68 €

Specifications

Color Green
Laboratory/Cleanroom Use Yes
Material Neoprene
Product Type Gloves
Style 25-101
View More Specs

Products 4
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
11781573
View Documents
Ansell™
25101100
10
30.68 €
Pack of 100
In Stock
to see stock.
Edge
Add to basket
 
11761573
View Documents
Ansell™
25101080
8
30.68 €
Pack of 100
In Stock
to see stock.
Edge
Add to basket
 
11771573
View Documents
Ansell™
25101090
9
29.50 €
Pack of 100
In Stock
to see stock.
Edge
Add to basket
 
11722125
View Documents
Ansell™
25101070
7
29.50 €
Pack of 100
In Stock
to see stock.
Edge
Add to basket
 
Description

Description

  • Polyurethane inner coating for easy donning
  • Textured fingertips provide a secure grip
  • Beaded cuff for secure hold
  • Green color
  • Highly resistant to acids, bases and alcohols
  • Good chemical splash resistance
Specifications

Specifications

Green
Neoprene
25-101
Beaded/Rolled
Acids, bases and alcohols
9.5 in.
Yes
Gloves
0.13 mm
Polymer
25-101
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ansell™ NeoTouch™ 25-101 Series Bright Green Neoprene Gloves

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.