missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nessler'S Solution, Pure, for Detection and Determination Of NH3 and Salts, Fisher Chemical™
Click to view available options
Quantity:
500 mL
1 L
Specifications
Specifications
Color | Yellow |
Packaging | Plastic Coated Glass bottle |
pH | 13 |
Quantity | 500 mL |
Physical Form | Liquid |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 11
Documents
Scientific Resources
Brochures
Fisherbrand Focus on pH and electrochemistry
File size: 9.1 MB
Fisherbrand Focus on pH and electrochemistry
File size: 2.5 MB
Safety and Handling




Product Identifier
- Nessler''s solution
- Danger
- Substances and mixtures corrosive to metals Category 1
- Acute toxicity Category 3
- Acute toxicity Category 2
- Acute toxicity Category 4
- Skin corrosion/irritation Category 1 A
- Serious eye damage/eye irritation Category 1
- Specific target organ toxicity after repeated exposure Category 2
- LONG-TERM AQUATIC HAZARD Chronic 2
- H290-May be corrosive to metals.
- H301-Toxic if swallowed.
- H310-Fatal in contact with skin.
- H332-Harmful if inhaled.
- H314-Causes severe skin burns and eye damage.
- H373-May cause damage to organs.
- H411-Toxic to aquatic life with long lasting effects.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/A§.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Nessler'S Solution, Pure, for Detection and Determination Of NH3 and Salts, Fisher Chemical™ >
Spot an opportunity for improvement?Share a Content Correction