Learn More
NeutraKine™VEGF165/121 Mouse anti-Human, Clone: 6C3D5, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 69025-1-IG-100UG
 View more versions of this product
Product Code. 17791199
  • 319.00 € / 100µg
Explore more special offers
Description
Description
NeutraKine®VEGF165/121 Monoclonal antibody specifically detects NeutraKine®VEGF165/121 in Human samples. It is validated for Neutralization, ELISA
Specifications
Specifications
NeutraKine™VEGF165/121
Monoclonal
Unconjugated
BC010129
Protein G purification
Primary
Human
Antibody
IgG1
Neutralization, ELISA
6C3D5
Sterile PBS.
Mouse
100 μg
7422
Store at -20°C