missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NeutraKine™VEGF165/121 Mouse anti-Human, Clone: 6C3D5, Proteintech
Mouse Monoclonal Antibody
149.00 € - 319.00 €
Specifications
Antigen | NeutraKine™VEGF165/121 |
---|---|
Clone | 6C3D5 |
Applications | Neutralization, ELISA |
Classification | Monoclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17701209
|
Proteintech
69025-1-IG-25UG |
25 μg |
149.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17791199
|
Proteintech
69025-1-IG-100UG |
100 μg |
319.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NeutraKine®VEGF165/121 Monoclonal antibody specifically detects NeutraKine®VEGF165/121 in Human samples. It is validated for Neutralization, ELISASpecifications
NeutraKine™VEGF165/121 | |
Neutralization, ELISA | |
Unconjugated | |
Mouse | |
Sterile PBS. | |
IgG1 | |
Protein G purification | |
Antibody |
6C3D5 | |
Monoclonal | |
Liquid | |
Human | |
7422 | |
Primary | |
Store at -20°C |