missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Neutral Red

78.89 € - 238.19 €

Chemical Identifiers

CAS 553-24-2
Molecular Formula C15H17ClN4
Molecular Weight (g/mol) 288.779
MDL Number MFCD00012651
InChI Key PGSADBUBUOPOJS-UHFFFAOYSA-N
Synonym Basic Red 5, C.I. 50040, 3-Amino-7-dimethylamino-2-methylphenazine hydrochloride
PubChem CID 11105
ChEBI CHEBI:86370
IUPAC Name 8-N,8-N,3-trimethylphenazine-2,8-diamine;hydrochloride
SMILES CC1=CC2=NC3=C(C=C(C=C3)N(C)C)N=C2C=C1N.Cl
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10259550
View Documents
Thermo Scientific Chemicals
415490250
25 g Glass bottle
78.89 €
25g
In Stock
to see stock.
Add to basket
 
10268473
View Documents
Thermo Scientific Chemicals
415491000
100 g Glass bottle
238.19 €
100g
In Stock
to see stock.
Add to basket
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
Specifications

Chemical Identifiers

553-24-2
288.779
PGSADBUBUOPOJS-UHFFFAOYSA-N
11105
8-N,8-N,3-trimethylphenazine-2,8-diamine;hydrochloride
C15H17ClN4
MFCD00012651
Basic Red 5, C.I. 50040, 3-Amino-7-dimethylamino-2-methylphenazine hydrochloride
CHEBI:86370
CC1=CC2=NC3=C(C=C(C=C3)N(C)C)N=C2C=C1N.Cl

Specifications

290.0°C
1 to 1.12
C15H17ClN4
25, 401
Basic Red 5, C.I. 50040, 3-Amino-7-dimethylamino-2-methylphenazine hydrochloride
PGSADBUBUOPOJS-UHFFFAOYSA-N
CC1=CC2=NC3=C(C=C(C=C3)N(C)C)N=C2C=C1N.Cl
288.779
CHEBI:86370
288.77
Green
Neutral Red, Pure
553-24-2
50% min.
MFCD00012651
15, 6573
Solubility in water: 50g/L. Other solubilities: 1.8% in abs. alcohol, 3.75% in cellosolve, 3.0% in ethylene glycol, practically insoluble in xylene
Authentic
8-N,8-N,3-trimethylphenazine-2,8-diamine;hydrochloride
11105
539 to 544nm
Pure
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Suspected of causing genetic defects.

GHS P Statement:
Use personal protective equipment as required.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge

Product Identifier
  • Neutral Red

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.