missing translation for 'onlineSavingsMsg'
Learn More
Ngamma-Trityl-L-asparagine hydrate, 98%, Thermo Scientific Chemicals Product Code.: 15429908

Ngamma-Trityl-L-asparagine hydrate, 98%, Thermo Scientific Chemicals

Product Code. 15429908
5 g
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy

Product Code. 15429908

Brand: Thermo Scientific Alfa Aesar H63285.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 132388-58-0
Molecular Formula C23H22N2O3
Molecular Weight (g/mol) 374.44
MDL Number MFCD00153422
InChI Key BRRPJQYCERAMFI-GNLPSFAGNA-N
Synonym h-asn trt-oh, n'-trityl-l-asparagine, n-beta-trityl-l-asparagine, n-triphenylmethyl-l-asparagine, 2s-2-amino-4-oxo-4-tritylamino butanoic acid, 2s-2-amino-3-triphenylmethylcarbamoyl propanoic acid, 2s-2-amino-3-triphenylmethyl carbamoyl propanoic acid, h-asn trt-2-chlorotrityl resin, asparagine trt, h-asn trt ohShow MoreShow Less
PubChem CID 7408309
IUPAC Name (2S)-2-amino-4-oxo-4-(tritylamino)butanoic acid
SMILES N[C@@H](CC(=O)NC(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1)C(O)=O

Specifications

Color White
Quantity 5 g
Formula Weight 392.45 (Anhydrous)
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Ngamma-Trityl-L-asparagine hydrate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Catalog Number
Lot Number5003I11P Certificate TypeCertificate of Analysis Date29/12/2023 Catalog NumberH63285.06,H63285.03
Lot Number81302258 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH63285.06,H63285.03

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ngamma-Trityl-L-asparagine hydrate, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.