Learn More
Nickel 2-methoxyethoxide, 5% w/v in 2-methoxyethanol, Thermo Scientific™
88.00 € - 279.00 €
Chemical Identifiers
CAS | 142600-62-2 |
---|---|
Molecular Formula | C6H14NiO4 |
Molecular Weight (g/mol) | 208.867 |
MDL Number | MFCD01863472 |
InChI Key | KNEAYFOSQJIPKZ-UHFFFAOYSA-N |
Synonym | nickel 2+ bis 2-methoxyethanolate |
PubChem CID | 131873637 |
IUPAC Name | 2-methoxyethanolate;nickel(2+) |
SMILES | COCC[O-].COCC[O-].[Ni+2] |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
142600-62-2 | |
208.867 | |
KNEAYFOSQJIPKZ-UHFFFAOYSA-N | |
131873637 | |
COCC[O-].COCC[O-].[Ni+2] |
C6H14NiO4 | |
MFCD01863472 | |
nickel 2+ bis 2-methoxyethanolate | |
2-methoxyethanolate;nickel(2+) |
Specifications
142600-62-2 | |
Ni(OCH2CH2OCH3)2 | |
UN1188 | |
nickel 2+ bis 2-methoxyethanolate | |
COCC[O-].COCC[O-].[Ni+2] | |
208.867 | |
208.76 | |
Liquid |
C6H14NiO4 | |
MFCD01863472 | |
Moisture sensitive | |
KNEAYFOSQJIPKZ-UHFFFAOYSA-N | |
2-methoxyethanolate;nickel(2+) | |
131873637 | |
5% w/v in 2-methoxyethanol | |
Nickel 2-methoxyethoxide |
Safety and Handling
GHS H Statement
H360-H351-H226-H302-H312-H332-H317
May damage fertility or the unborn child.
Suspected of causing cancer.
Flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
P201-P202-P210-P233-P240-P241-P242-P243-P260-P264b-P270-P271-P272-P280g-P281-P301+P312-P303+P361+P353-P304+P340-P307+P311-P312-P330-P333+P313-P363-P370+P378q-P501c
H226-H302+H312+H332-H317-H350-H360FD-H370-H373
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ETHYLENE GLYCOL MONOMETHYL ETHER
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only