missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nickel Electro™ Nickel 201 Dish Lids
Nickel Electro™ Nickel 201 Dish Lids are made to resist corrosive enviroments, which make it useful for maintaining product purity.
Brand: Nickel Electro™ 546
Description
- 99.6% nickel purity
- Crevice free
- High thermal and electrical conductivities
- Low gas content
- Low vapor pressure
- Ideal for lab crucibles that are capable of withstanding oxidizing furnace atmospheres up to 1100°C
Specifications
Nickel | |
Dish Lid | |
85 mm |