missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nickel Electro™ Nickel 201 Dish Lids
Nickel Electro™ Nickel 201 Dish Lids are made to resist corrosive enviroments, which make it useful for maintaining product purity.
20.20 € - 25.25 €
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | |||||
12747376
|
Nickel Electro™
543 |
60 mm |
21.70 €
Each |
Estimated Shipment: 02-01-2025
Log in to see stock. |
|||||
12787376
|
Nickel Electro™
548 |
90 mm |
23.60 €
Each |
Estimated Shipment: 02-01-2025
Log in to see stock. |
|||||
12777376
|
Nickel Electro™
546 |
85 mm |
25.25 €
Each |
Estimated Shipment: 18-12-2024
Log in to see stock. |
|||||
12737376
|
Nickel Electro™
541 |
55 mm |
20.20 €
Each |
Estimated Shipment: 02-01-2025
Log in to see stock. |
|||||
12757376
|
Nickel Electro™
544 |
70 mm |
22.35 €
Each |
Estimated Shipment: 02-01-2025
Log in to see stock. |
|||||
12767376
|
Nickel Electro™
545 |
75 mm |
20.60 €
Each |
Estimated Shipment: 02-01-2025
Log in to see stock. |
|||||
Description
- 99.6% nickel purity
- Crevice free
- High thermal and electrical conductivities
- Low gas content
- Low vapor pressure
- Ideal for lab crucibles that are capable of withstanding oxidizing furnace atmospheres up to 1100°C
Specifications
Nickel | |
Dish Lid |