Learn More
Nickel foil, 2.0mm (0.08in) thick, 99.5% (metals basis), Thermo Scientific Chemicals
59.30 € - 671.34 €
Chemical Identifiers
CAS | 7440-02-0 |
---|---|
Molecular Formula | Ni |
Molecular Weight (g/mol) | 58.69 |
MDL Number | MFCD00011137 MFCD06798735 |
InChI Key | PXHVJJICTQNCMI-UHFFFAOYSA-N |
Synonym | raney alloy, fibrex, catalyst, particles, fibrex p, nickel, elemental, nichel italian, nickel, soluble salts, carbonyl powder, niccolum |
PubChem CID | 935 |
ChEBI | CHEBI:28112 |
IUPAC Name | nickel |
SMILES | [Ni] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11386219
|
Thermo Scientific Alfa Aesar
044825.FI |
50 x 50 mm |
59.30 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11396219
|
Thermo Scientific Alfa Aesar
044825.GH |
100 x 100 mm |
158.79 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11306229
|
Thermo Scientific Alfa Aesar
044825.HP |
100 x 500 mm |
671.34 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7440-02-0 | |
58.69 | |
PXHVJJICTQNCMI-UHFFFAOYSA-N | |
935 | |
nickel |
Ni | |
MFCD00011137 MFCD06798735 | |
raney alloy, fibrex, catalyst, particles, fibrex p, nickel, elemental, nichel italian, nickel, soluble salts, carbonyl powder, niccolum | |
CHEBI:28112 | |
[Ni] |
Specifications
7440-02-0 | |
MFCD00011137 MFCD06798735 | |
PXHVJJICTQNCMI-UHFFFAOYSA-N | |
nickel | |
935 | |
99.5% | |
≈44.5g/50x50mm | |
(metals basis) |
Ni | |
raney alloy, fibrex, catalyst, particles, fibrex p, nickel, elemental, nichel italian, nickel, soluble salts, carbonyl powder, niccolum | |
[Ni] | |
58.69 | |
CHEBI:28112 | |
Odorless | |
Foil | |
Nickel foil, 2.0mm (0.08 in.) thick |
Safety and Handling
GHS H Statement
H372-H351-H317
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing cancer.
May cause an allergic skin reaction.
P201-P202-P260-P261-P264b-P270-P272-P280-P302+P352-P308+P313-P333+P313-P363-P501c
H317-H351-H372
missing translation for 'einecsNumber' : 231-111-4
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.