Learn More
Nickel(II) acetate tetrahydrate, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals
106.53 € - 454.26 €
Chemical Identifiers
CAS | 6018-89-9 |
---|---|
Molecular Formula | C4H14NiO8 |
Molecular Weight (g/mol) | 248.84 |
MDL Number | MFCD00066973 |
InChI Key | OINIXPNQKAZCRL-UHFFFAOYSA-L |
Synonym | nickel ii acetate tetrahydrate, nickel acetate tetrahydrate, nickel diacetate tetrahydrate, diacetatonickel tetrahydrate, nickelous acetate tetrahydrate, unii-6soa8l0560, acetic acid, nickel +2 salt, tetrahydrate, nickel 2+ diacetate tetrahydrate, nickel acetate tetrahydrate nickel and nickel compounds, acetic acid, nickel 2+ salt, tetrahydrate |
PubChem CID | 62601 |
IUPAC Name | nickel(2+);diacetate;tetrahydrate |
SMILES | O.O.O.O.[Ni++].CC([O-])=O.CC([O-])=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11306937
|
Thermo Scientific Alfa Aesar
010813.06 |
5 g |
106.53 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11396927
|
Thermo Scientific Alfa Aesar
010813.14 |
25 g |
454.26 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It is used in electroplating, anodize coatings; textile dyeing mordant; Catalyst. Also, used in synthesis of polynuclear-nickel polyoxotungstate cluster compounds that are ideal for the design of molecular magnets.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
6018-89-9 | |
248.84 | |
OINIXPNQKAZCRL-UHFFFAOYSA-L | |
62601 | |
O.O.O.O.[Ni++].CC([O-])=O.CC([O-])=O |
C4H14NiO8 | |
MFCD00066973 | |
nickel ii acetate tetrahydrate, nickel acetate tetrahydrate, nickel diacetate tetrahydrate, diacetatonickel tetrahydrate, nickelous acetate tetrahydrate, unii-6soa8l0560, acetic acid, nickel +2 salt, tetrahydrate, nickel 2+ diacetate tetrahydrate, nickel acetate tetrahydrate nickel and nickel compounds, acetic acid, nickel 2+ salt, tetrahydrate | |
nickel(2+);diacetate;tetrahydrate |
Specifications
(decomposition) | |
Green | |
C4H14NiO8 | |
MFCD00066973 | |
3730764 | |
nickel ii acetate tetrahydrate, nickel acetate tetrahydrate, nickel diacetate tetrahydrate, diacetatonickel tetrahydrate, nickelous acetate tetrahydrate, unii-6soa8l0560, acetic acid, nickel +2 salt, tetrahydrate, nickel 2+ diacetate tetrahydrate, nickel acetate tetrahydrate nickel and nickel compounds, acetic acid, nickel 2+ salt, tetrahydrate | |
OINIXPNQKAZCRL-UHFFFAOYSA-L | |
nickel(2+);diacetate;tetrahydrate | |
62601 | |
99.999% | |
Light | |
(metals basis) |
6018-89-9 | |
Crystalline Aggregates | |
Ni(CH3COO)2·4H2O | |
UN3077 | |
14,6499 | |
Soluble in water and methanol. Insoluble in diethyl ether,n-octanol. | |
O.O.O.O.[Ni++].CC([O-])=O.CC([O-])=O | |
248.84 | |
248.84 (176.79 Anhydrous) | |
Puratronic™ | |
1.744 g/mL | |
Nickel(II) acetate tetrahydrate |
Safety and Handling
GHS H Statement
H334-H350-H360-H372-H341-H302-H332-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause cancer.
May damage fertility or the unborn child.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing genetic defects.
Harmful if swallowed.
Harmful if inhaled.
May cause an allergic skin reaction.
P201-P202-P260-P264b-P270-P271-P272-P280g-P281-P285-P301+P312-P302+P352-P304+P340-P308+P313-P330-P333+P313-P342+P311-P363-P501c
H302+H332-H317-H334-H335-H341-H350i-H360D-H372
missing translation for 'dotInformation' : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
missing translation for 'einecsNumber' : 206-761-7
missing translation for 'rtecsNumber' : QR6126000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.