missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nickel(II) bromide, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 963.0°C |
Color | Brown |
CAS Min % | 98.5 |
CAS Max % | 100.0 |
Physical Form | Crystalline Powder and/or Chunks |
Quantity | 250 g |
Assay Percent Range | 98.5% min. (on dry substance) (Argentometry) |
Linear Formula | NiBr2 |
Fieser | 04,351 |
Merck Index | 15, 6587 |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 6
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0466508 | Certificate of Analysis | 02/10/2024 | 203362500,203360100,203360500 |
A0449916 | Certificate of Analysis | 02/05/2023 | 203362500,203360100,203360500 |
A0403396 | Certificate of Analysis | 15/12/2022 | 203362500,203360100,203360500 |
A0409518 | Certificate of Analysis | 13/12/2022 | 203362500,203360100,203360500 |
A0418398 | Certificate of Analysis | 12/12/2022 | 203362500,203360100,203360500 |
Safety and Handling


Product Identifier
- Nickel(II) bromide
- Danger
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Carcinogenicity Category 1A
- Germ cell mutagenicity Category 2
- Reproductive toxicity Category 1B
- Respiratory sensitiser Category 1
- Skin sensitiser Category 1
- Specific target organ toxicity Category 1
- H317-May cause an allergic skin reaction.
- H372-Causes damage to organs.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H350i-May cause cancer by inhalation.
- H360D-May damage the unborn child.
- H341-Suspected of causing genetic defects.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- P201-Obtain special instructions before use.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Nickel(II) bromide, 99%, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction