Learn More
Nickel(II) carbonate (basic) hydrate, Ni 40% min, typically 99.5% (metals basis), Thermo Scientific Chemicals
Nickel Plating pH Adjuster, Catalysts, Ceramic Colors and Glazes
61.81 € - 134.67 €
Chemical Identifiers
CAS | 155775-31-8 |
---|---|
Molecular Formula | CH4Ni3O7 |
Molecular Weight (g/mol) | 304.12 |
MDL Number | MFCD00016262 |
InChI Key | ATYNVYNPWGEMMI-UHFFFAOYSA-H |
Synonym | nickel 3+ ;carbonate;hydroxide;hydrate, nickel carbonate, basic hydrate trace metals basis |
PubChem CID | 86176052 |
IUPAC Name | trinickel(2+) tetrahydroxide carbonate |
SMILES | [OH-].[OH-].[OH-].[OH-].[Ni++].[Ni++].[Ni++].[O-]C([O-])=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11369237
|
Thermo Scientific Alfa Aesar
040106.30 |
250 g |
61.81 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11359237
|
Thermo Scientific Alfa Aesar
040106.A1 |
1 kg |
134.67 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Nickel plating pH adjuster, catalysts, ceramic colors and glazes.Nickel(II) carbonate (basic) hydrate is used as a precursor to catalysts and in ceramic applications. It acts as an intermediate in the hydrometallurgical purification of nickel from its ores. It is also used in electroplating of nickel.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
155775-31-8 | |
304.12 | |
ATYNVYNPWGEMMI-UHFFFAOYSA-H | |
86176052 | |
[OH-].[OH-].[OH-].[OH-].[Ni++].[Ni++].[Ni++].[O-]C([O-])=O |
CH4Ni3O7 | |
MFCD00016262 | |
nickel 3+ ;carbonate;hydroxide;hydrate, nickel carbonate, basic hydrate trace metals basis | |
trinickel(2+) tetrahydroxide carbonate |
Specificatie
155775-31-8 | |
Powder | |
CH4Ni3O7 | |
nickel 3+ ;carbonate;hydroxide;hydrate, nickel carbonate, basic hydrate trace metals basis | |
ATYNVYNPWGEMMI-UHFFFAOYSA-H | |
trinickel(2+) tetrahydroxide carbonate | |
86176052 | |
Odorless | |
Nickel(II) carbonate (basic) hydrate |
Green | |
99.5% (metals basis) | |
MFCD00016262 | |
Insoluble in water. | |
[OH-].[OH-].[OH-].[OH-].[Ni++].[Ni++].[Ni++].[O-]C([O-])=O | |
304.12 | |
304.18 (Anhydrous) | |
Green powder |
Veiligheid en behandeling
P201-P202-P260-P264b-P270-P271-P272-P280-P281-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P330-P333+P313-P342+P311-P362-P501c
H302+H332-H315-H317-H319-H334-H341-H350i-H360D-H372
missing translation for 'dotInformation' : DOT Class: 9, Packaging Group: III
missing translation for 'einecsNumber' : 240-408-8
missing translation for 'rtecsNumber' : QR6260000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.