missing translation for 'onlineSavingsMsg'
Learn More

Nickel(II) perchlorate hexahydrate, Reagent Grade, Thermo Scientific Chemicals

32.46 € - 241.20 €

Chemical Identifiers

CAS 13520-61-1
Molecular Formula Cl2H12NiO14
Molecular Weight (g/mol) 365.675
MDL Number MFCD00149804
InChI Key WHGYCGOFTBFDLW-UHFFFAOYSA-L
Synonym nickel ii perchlorate hexahydrate, acmc-1boln, 2clo4.ni.6h2o, nickel ii perchlorate hydrate, nickel 2+ hexahydrate diperchlorate, nickel perchlorate, hexahydrate, reagent, nickel ii perchlorate hexahydrate, reagent grade, nickel ii perchlorate hexahydrate trace metals basis, nickel ii perchlorate hexahydrate, purum p.a., crystallized ktShow MoreShow Less
PubChem CID 15769463
IUPAC Name nickel(2+);diperchlorate;hexahydrate
SMILES O.O.O.O.O.O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Ni+2]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11391728
SDS View Documents
Thermo Scientific Alfa Aesar
011631.14
25 g
32.46 €
25g
Please to purchase this item. Need a web account? Register with us today!
11381728
SDS View Documents
Thermo Scientific Alfa Aesar
011631.22
100 g
74.47 €
100g
Please to purchase this item. Need a web account? Register with us today!
11371728
SDS View Documents
Thermo Scientific Alfa Aesar
011631.36
500 g
241.20 €
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Nickel(II) perchlorate hexahydrate is used as a starting material for homometallic, trinuclear heteroscorpionate complexes applied in studies of electronic and magnetic properties.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling
Show More
SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Nickel(II) perchlorate hexahydrate, Reagent Grade, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.