Learn More
Nickel Iron Molybdenum flake, -200 mesh, Thermo Scientific Chemicals
Brand: Thermo Scientific Alfa Aesar 045610.30
107.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Nickel Iron Molybdenum flake | |
0.29g/mL | |
UN3089 | |
−200 Mesh |
Ni:Fe:Mo; 81:17:2 wt% | |
MFCD09910537 | |
250 g | |
Flake |
Safety and Handling
- Nickel Iron Molybdenum flake, -200 mesh
Signal Word
- Danger
Hazard Category
- Flammable solid Category 2
- Serious eye damage/eye irritation Category 2
- Skin sensitiser Category 1
- Carcinogenicity Category 2
- Specific target organ toxicity Category 1
Hazard Statement
- H228-Flammable solid.
- H319-Causes serious eye irritation.
- H317-May cause an allergic skin reaction.
- H351-Suspected of causing cancer.
- H372-Causes damage to organs.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501-Dispose of contents/container to .
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
RUO – Research Use Only