missing translation for 'onlineSavingsMsg'
Learn More

Nickel Iron Molybdenum flake, -200 mesh, Thermo Scientific Chemicals

107.54 € - 244.22 €

Specifications

Chemical Name or Material Nickel Iron Molybdenum flake
Avg. Mol. Wt. or Mol. Wt. Range Ni:Fe:Mo; 81:17:2 wt%
Density 0.29g/mL
MDL Number MFCD09910537
UN Number UN3089
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15412417
View Documents
Thermo Scientific Alfa Aesar
045610.30
250 g
107.54 €
250g
Please to purchase this item. Need a web account? Register with us today!
15422417
View Documents
Thermo Scientific Alfa Aesar
045610.A1
1 kg
244.22 €
1kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Nickel Iron Molybdenum flake
0.29g/mL
UN3089
Flake
Ni:Fe:Mo; 81:17:2 wt%
MFCD09910537
−200 Mesh
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H228-H351-H317
Flammable solid.
Suspected of causing cancer.
May cause an allergic skin reaction.

P201-P202-P210-P240-P241-P260-P264b-P270-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P370+P378q-P501c

H228-H317-H350-H372

missing translation for 'dotInformation' : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: METAL POWDERS, FLAMMABLE, N.O.S.

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Product Identifier
  • Nickel Iron Molybdenum flake, -200 mesh

Signal Word
  • Danger

Hazard Category
  • Flammable solid Category 2
  • Serious eye damage/eye irritation Category 2
  • Skin sensitiser Category 1
  • Carcinogenicity Category 2
  • Specific target organ toxicity Category 1

Hazard Statement
  • H228-Flammable solid.
  • H319-Causes serious eye irritation.
  • H317-May cause an allergic skin reaction.
  • H351-Suspected of causing cancer.
  • H372-Causes damage to organs.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.