Learn More
Nickel plating solution, electroless, ammonia free, Thermo Scientific Chemicals
For plating on silicon, germainium, gallium arsenide, cadmium sulfide, nickel or Kovar
Brand:  Thermo Scientific Alfa Aesar 044070.K2
 View more versions of this product
Product Code. 11395148
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
For plating on silicon, germainium, gallium arsenide, cadmium sulfide, nickel or Kovar
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Nickel plating solution
Electroless, ammonia free
UN3082
Green
MFCD00011137
1 L