missing translation for 'onlineSavingsMsg'
Learn More

Nickel plating solution, electroless, ammonia free, Thermo Scientific Chemicals

For plating on silicon, germainium, gallium arsenide, cadmium sulfide, nickel or Kovar

27.30 € - 79.90 €

Specifications

Chemical Name or Material Nickel plating solution
Color Green
Name Note Electroless, ammonia free
MDL Number MFCD00011137
UN Number UN3082
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11305158
View Documents Promotion Details
Thermo Scientific Alfa Aesar
044070.AK
250 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
11395148
View Documents Promotion Details
Thermo Scientific Alfa Aesar
044070.K2
1 L This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

For plating on silicon, germainium, gallium arsenide, cadmium sulfide, nickel or Kovar

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Nickel plating solution
Electroless, ammonia free
UN3082
Green
MFCD00011137
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H334-H350-H360-H372-H341-H302-H332-H315-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause cancer.
May damage fertility or the unborn child.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing genetic defects.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
May cause an allergic skin reaction.

GHS P Statement
P273-P201-P309+P311-P501a
Avoid release to the environment.
Obtain special instructions before use.
IF exposed or if you feel unwell: Call a POISON CENTER or doctor/physician.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

missing translation for 'dotInformation' : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.

missing translation for 'tsca' : TSCA

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only