Learn More
Nickel sulfide, 99.9% (metals basis), Thermo Scientific Chemicals
166.83 € - 1778.00 €
Chemical Identifiers
CAS | 12035-72-2 |
---|---|
Molecular Formula | Ni3S2 |
Molecular Weight (g/mol) | 240.20 |
MDL Number | MFCD01711880 |
InChI Key | YGHCWPXPAHSSNA-UHFFFAOYSA-N |
Synonym | nickel subsulfide, trinickel disulphide, nickel sulfide, trinickel disulfide, nickel subsulphide, nickel sesquisulfide, nickel tritadisulphide, nickel iii sulfide, alpha nickel subsulfide, ccris 432 |
PubChem CID | 22833473 |
SMILES | S=[Ni][Ni][Ni]=S |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11485040
|
Thermo Scientific Alfa Aesar
035661.09 |
10 g |
166.83 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11383807
|
Thermo Scientific Alfa Aesar
035661.18 |
50 g |
436.17 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11393807
|
Thermo Scientific Alfa Aesar
035661.30 |
250 g | N/A | ||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
12035-72-2 | |
240.20 | |
YGHCWPXPAHSSNA-UHFFFAOYSA-N | |
22833473 |
Ni3S2 | |
MFCD01711880 | |
nickel subsulfide, trinickel disulphide, nickel sulfide, trinickel disulfide, nickel subsulphide, nickel sesquisulfide, nickel tritadisulphide, nickel iii sulfide, alpha nickel subsulfide, ccris 432 | |
S=[Ni][Ni][Ni]=S |
Specifications
797°C | |
Black | |
99.9% (metals basis) | |
MFCD01711880 | |
Soluble in water(0,00735g/L at 20°C). | |
S=[Ni][Ni][Ni]=S | |
240.20 | |
240.19 | |
Nickel sulfide |
12035-72-2 | |
Powder | |
Ni3S2 | |
nickel subsulfide, trinickel disulphide, nickel sulfide, trinickel disulfide, nickel subsulphide, nickel sesquisulfide, nickel tritadisulphide, nickel iii sulfide, alpha nickel subsulfide, ccris 432 | |
YGHCWPXPAHSSNA-UHFFFAOYSA-N | |
disulfanylidenetrinickel | |
22833473 | |
Black powder |
Safety and Handling
P201-P202-P260-P264b-P270-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c
H317-H341-H350i-H372
missing translation for 'dotInformation' : DOT Class: 9; Packing Group: III
missing translation for 'einecsNumber' : 234-829-6
missing translation for 'rtecsNumber' : QR9800000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only