missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Nile Red 99%

134.67 € - 863.30 €

Chemical Identifiers

CAS 7385-67-3
Molecular Formula C20H18N2O2
Molecular Weight (g/mol) 318.38
MDL Number MFCD00011639
InChI Key VOFUROIFQGPCGE-UHFFFAOYSA-N
Synonym Nile blue A oxazone
PubChem CID 65182
ChEBI CHEBI:52169
SMILES CCN(CC)C1=CC=C2N=C3C(OC2=C1)=CC(=O)C1=CC=CC=C31
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10464311
View Documents
Thermo Scientific Chemicals
415711000
100 mg Glass bottle
134.67 €
100mg
In Stock
to see stock.
Add to basket
 
10658904
View Documents
Thermo Scientific Chemicals
415710010
1 g Glass bottle
863.30 €
1g
In Stock
to see stock.
Add to basket
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7385-67-3
318.38
VOFUROIFQGPCGE-UHFFFAOYSA-N
65182
CCN(CC)C1=CC=C2N=C3C(OC2=C1)=CC(=O)C1=CC=CC=C31
C20H18N2O2
MFCD00011639
Nile blue A oxazone
CHEBI:52169

Specifications

203.0°C to 205.0°C
98.5
99%
MFCD00011639
15, 6628
Solubility in water: almost insoluble. Other solubilities: soluble in acetone, acetonitrile, chloroform, thf,, ethanol, dmso, cs2, benzene, nitrobenzene, pyridin, e, hexane, diethyl ether
Authentic
9-(diethylamino)benzo[a]phenoxazin-5-one
65182
318.38
Pure
Complies
Powder
7385-67-3
100.0
C20H18N2O2
27, II, 468
Nile blue A oxazone
VOFUROIFQGPCGE-UHFFFAOYSA-N
CCN(CC)C1=CC=C2N=C3C(OC2=C1)=CC(=O)C1=CC=CC=C31
318.38
CHEBI:52169
99%
98.5% min.
Maroon to Red
Nile Red, 99%
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 230-966-0

missing translation for 'tsca' : TSCA

Product Identifier
  • Nile Red

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.