missing translation for 'onlineSavingsMsg'
Learn More

Niobium(IV) oxide, 99+% (metals basis), Thermo Scientific Chemicals

Chemical Identifiers

CAS 12034-59-2
Molecular Formula NbO2
Molecular Weight (g/mol) 124.90
MDL Number MFCD00016242
InChI Key HFLAMWCKUFHSAZ-UHFFFAOYSA-N
Synonym niobium dioxide, niobium iv oxide, niobium oxide, columbium dioxide, niobium iv oxide trace metals basis, niobium iv oxide trace metals basis 10g
PubChem CID 82839
IUPAC Name dioxoniobium
SMILES O=[Nb]=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11493810
View Documents Promotion Details
Thermo Scientific Alfa Aesar
089692.09
10 g N/A
N/A
11483810
View Documents Promotion Details
Thermo Scientific Alfa Aesar
089692.18
50 g N/A
N/A
Description

Description

Niobium(IV) oxide is used as a reducing agent, which reduces carbon dioxide and sulfur dioxide to carbon and sulfur respectively. It finds application in dielectric material for electronic capacitors. It is also used for manufacturing metal niobium metal, in optical glass and in electronic industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

12034-59-2
124.90
HFLAMWCKUFHSAZ-UHFFFAOYSA-N
82839
O=[Nb]=O
NbO2
MFCD00016242
niobium dioxide, niobium iv oxide, niobium oxide, columbium dioxide, niobium iv oxide trace metals basis, niobium iv oxide trace metals basis 10g
dioxoniobium

Specifications

12034-59-2
>99% (metals basis)
MFCD00016242
Insoluble in water.
O=[Nb]=O
124.90
124.91
Niobium(IV) oxide
Powder
NbO2
niobium dioxide, niobium iv oxide, niobium oxide, columbium dioxide, niobium iv oxide trace metals basis, niobium iv oxide trace metals basis 10g
HFLAMWCKUFHSAZ-UHFFFAOYSA-N
dioxoniobium
82839
5.9 g/mL
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 234-809-7

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only