missing translation for 'onlineSavingsMsg'
Learn More
Niobium(V) ethoxide, 99.99% (metals basis), Ta <500ppm, Thermo Scientific Chemicals Product Code.: 11376998

Niobium(V) ethoxide, 99.99% (metals basis), Ta <500ppm, Thermo Scientific Chemicals

Product Code. 11376998
10 g
Click to view available options
Quantity:
2 g
10 g
50 g
This item is not returnable. View return policy

Product Code. 11376998

Brand: Thermo Scientific Alfa Aesar 014689.09

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Preparation of niobium oxide and mixed metal oxides

Preparation of niobium oxide and mixed metal oxides.Niobium(V) ethoxide is used for the sol-gel processing of materials containing niobium oxides and mixed metal oxides. It is also used to prepare ferroelectric nanomaterials consisting of oxides of potassium, silicon and niobium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3236-82-6
Molecular Formula C10H25NbO5
Molecular Weight (g/mol) 318.21
MDL Number MFCD00015122
InChI Key ZTILUDNICMILKJ-UHFFFAOYSA-N
Synonym niobium ethoxide, niobium 5+ ethanolate, ethanol, niobium 5+ salt, niobium 5+ ion pentakis ethoxide, pentaethoxyniobium, niobium pentaethoxide, ethanol, niobium 5+ salt 5:1, acmc-1cmht, niobium v pentaethoxide, ethanolate; niobium 5+Show MoreShow Less
PubChem CID 160675
IUPAC Name ethanolate;niobium(5+)
SMILES CCO[Nb](OCC)(OCC)(OCC)OCC

Specifications

Melting Point 6°C
Density 1.258 g/mL
Boiling Point 140°C to 142°C (0.1 mmHg)
Flash Point 36°C (97°F)
Linear Formula Nb(OC2H5)5
Refractive Index 1.516
Quantity 10 g
UN Number UN2924
Beilstein 3678883
Sensitivity Moisture sensitive
Solubility Information Soluble in organic solvents; Decomposes in water;Miscible with organic solvents.
Formula Weight 318.21
Percent Purity 99.999%
Concentration or Composition (by Analyte or Components) Ta <500ppm
Physical Form Liquid
Assay (metals basis)
Chemical Name or Material Niobium(V) ethoxide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberT11K035 Certificate TypeCertificate of Analysis Date26/07/2024 Catalog Number014689.09,014689.18,014689.04
Lot NumberP18K037 Certificate TypeCertificate of Analysis Date02/04/2024 Catalog Number014689.09,014689.18,014689.04
Lot NumberZ22J010 Certificate TypeCertificate of Analysis Date26/01/2024 Catalog Number014689.09,014689.18,014689.04
Lot NumberY10J009 Certificate TypeCertificate of Analysis Date01/01/2024 Catalog Number014689.09,014689.18,014689.04
Lot NumberX19H020 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number014689.09,014689.18,014689.04

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Niobium(V) ethoxide, 99.99% (metals basis), Ta <500ppm, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.