missing translation for 'onlineSavingsMsg'
Learn More
Titanium(IV) isopropoxide, 98+%, Thermo Scientific Chemicals Product Code.: 10792041

Titanium(IV) isopropoxide, 98+%, Thermo Scientific Chemicals

Product Code. 10792041
250 mL, Glass bottle
Click to view available options
Quantity:
5 mL
250 mL
1 L
2.5 L
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10792041

Brand: Thermo Scientific Acros 194702500

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Lewis acid
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 546-68-9
Molecular Formula C12H28O4Ti
Molecular Weight (g/mol) 284.26
InChI Key VXUYXOFXAQZZMF-UHFFFAOYSA-N
Synonym titanium tetraisopropanolate, titanium iv isopropoxide, tetraisopropyl orthotitanate, titanium isopropoxide, titanium tetraisopropylate, titanium isopropylate, ti isopropylate, tetraisopropoxytitanium iv, isopropyl orthotitanate, tetraisopropyl titanateShow MoreShow Less
PubChem CID 11026
IUPAC Name propan-2-olate;titanium(4+)
SMILES CC(C)[O-].CC(C)[O-].CC(C)[O-].CC(C)[O-].[Ti+4]

Specifications

Melting Point >15.0°C
Density 0.9500g/mL
Boiling Point 240.0°C
Flash Point 46°C
Infrared Spectrum Authentic
Assay Percent Range 16.6 to 17.3% (Ti)
Packaging Glass bottle
Linear Formula Ti[OCH(CH3)2]4
Refractive Index 1.4654 to 1.4684
Quantity 250 mL
Beilstein 01,II,382
Fieser 11,92; 12,90; 13,13; 14,61; 15,308; 16,54; 17,347
Merck Index 15,9636
Specific Gravity 0.95
Solubility Information Solubility in water: hydrolysis. Other solubilities: soluble in most common organic solvents
Viscosity 4.3 mPa.s (25°C)
Formula Weight 284.26
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material Titanium(IV) isopropoxide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0447598 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number194700010,194700050,194700025,194702500
Lot NumberA0468569 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number194700010,194700050,194700025,194702500
Lot NumberA0464708 Certificate TypeCertificate of Analysis Date10/09/2024 Catalog Number194700010,194700050,194700025,194702500
Lot NumberA0447598 Certificate TypeCertificate of Analysis Date12/12/2023 Catalog Number194700010,194700050,194700025,194702500
Lot NumberA0456982 Certificate TypeCertificate of Analysis Date17/10/2023 Catalog Number194700010,194700050,194700025,194702500
compliance-icons compliance-icons
Product Identifier
  • Titanium(IV) isopropoxide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H331-Toxic if inhaled.
  • H336-May cause drowsiness or dizziness.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Titanium(IV) isopropoxide, 98+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.