Learn More
Nitazoxanide-d4, MedChemExpress
MedChemExpress Nitazoxanide D4 (NTZ D4) is the deuterium labeled Nitazoxanide, which is an antiprotozoal agent.
Brand:  Medchem Express HY-B0217S
Encompass_Preferred
 View more versions of this product
Product Code. 16470828
  • POA / 5mg
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Explore more special offers
Description
Description
Nitazoxanide D4 (NTZ D4) is the deuterium labeled Nitazoxanide, which is an antiprotozoal agent.
Specifications
Specifications
Nitazoxanide-d4
5 mg
C12H5D4N3O5S
NTZ-d4 NSC-697855-d4
O=C(NC1=NC=C([N+]([O-])=O)S1)C2=C(OC(C)=O)C([2H])=C([2H])C([2H])=C2[2H]
311.31
Research
1246819-17-9
Research
Solid
Off-White
311.31
98.0%
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Nitazoxanide-d4, MedChemExpress