missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nitric Acid, 69.0 to 70.0%, N.F., J.T. Baker™
Click to view available options
Quantity:
2.5 L
Packaging:
Glass Bottle
pvc coated glass bottle
Specifications
Specifications
Packaging | Glass Bottle |
Specific Gravity | 1L = 1.41kg |
Quantity | 2.5 L |
Formula Weight | 63.01 |
Percent Purity | 69.0 to 70.0% |
Grade | NF |
Chemical Name or Material | Nitric Acid |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Safety and Handling


Signal Word
- Danger
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
- Oxidising liquid Category 2
- Skin corrosion/irritation Category 1 A
- H272-May intensify fire; oxidiser.
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P405-Store locked up.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Nitric Acid, 69.0 to 70.0%, N.F., J.T. Baker™ >
Spot an opportunity for improvement?Share a Content Correction