Learn More
Lenz Laborglasintrumente Nitrogen Tubes (Schlenk Tubes) with PTFE Needle-Valve Stopcock
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
94.00 € - 114.00 €
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
11923421
|
Lenz Laborglasintrumente
03370649 |
250 mL |
114.00 €
Each |
Estimated Shipment: 30-05-2025
Log in to see stock. |
||||||
11983411
|
Lenz Laborglasintrumente
03370613 |
10 mL |
94.00 €
Each |
Estimated Shipment: 04-06-2025
Log in to see stock. |
||||||
Description
- PTFE needle-valve stopcock
- With conical ground joint and PTFE stopcock
- Made of DURAN™ tubing
Specifications
Duran Glass |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Lenz Laborglasintrumente Nitrogen Tubes (Schlenk Tubes) with PTFE Needle-Valve Stopcock