Learn More
Lenz Laborglasintrumente Nitrogen Tubes (Schlenk Tubes) with PTFE Needle-Valve Stopcock
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand:  Lenz Laborglasintrumente 03370649
 View more versions of this product
Product Code. 11923421
  • 114.00 € / Each
Estimated Shipment: 11-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • PTFE needle-valve stopcock
  • With conical ground joint and PTFE stopcock
  • Made of DURAN™ tubing
Specifications
Specifications
250 mL
Duran Glass
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Lenz Laborglasintrumente Nitrogen Tubes (Schlenk Tubes) with PTFE Needle-Valve Stopcock > 250 mL