missing translation for 'onlineSavingsMsg'
Learn More

3M™ Nomad™ Terra Heavy Traffic Entrance Matting 8250

Nomad™ Terra Heavy Traffic Entrance Matting 8250 is coiled vinyl scraper matting suitable for use indoors, outdoors, or in recessed wells with high traffic volumes.

Brand:  3M™ 7100283949

960.93 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.


Product Code. 17823256

  • 1680.00 € / Each
Estimated Shipment: 28-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Trap and hold dirt and moisture in high traffic areas with 3M™ Nomad™ Terra Heavy Traffic Entrance Matting 8250.
  • Our coiled vinyl scraper matting is suitable for use indoors, outdoors, or in recessed wells in high traffic areas, offering excellent resistance to grease and oil.
  • The open coil structure removes, traps and hides dirt and moisture, preventing it from being tracked back into the building.
  • The cushioned vinyl backing protects floor surfaces in indoor areas and makes it easy to clean.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Dark Gray
6 m
Entrance Mat
1.2 m
19.685 ft.
Vinyl
3.937 ft.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3M™ Nomad™ Terra Heavy Traffic Entrance Matting 8250 > Entrance Matting

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.