missing translation for 'onlineSavingsMsg'
Learn More
Learn More
QIAGEN QIAcard FTA Classic
For collection, stabilization, processing, transport and archiving of biological samples. Non-Indicating QIAcard FTA Classic with 4 sample areas per card.
364.00 € - 920.00 €
Specifications
Description | Collect, stabilize, process, transport, and archive blood samples with Non-Indicating FTA Classic Cards. |
---|---|
Format | Card |
Sample Type | Blood |
For Use With (Equipment) | Direct STR amplification |
For Use With (Application) | Forensics, Transgenic identification, Transfusion medicine/HLA typing, Plasmid screening, Food and agriculture testing, Drug discovery, Genomics, STR analysis, Animal identification, Whole genome amplification, Molecular biology |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11310854
|
QIAGEN
WB120305 |
25 Pk. |
364.00 €
Pack of 25 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10223352
|
QIAGEN
WB120205 |
100 Pk. |
920.00 €
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
For collection, stabilization, processing, transport and archiving of biological samples. Non-Indicating QIAcard FTA Classic with 4 sample areas per card.
- QIAcard FTA Classic format with 4 spot area to collect, stabilize, process, transport and archive colored samples such as blood.
- FTA technology enables cell lysis on contact, denatures proteins and immediate stabilization and protection of nucleic acids.
Specifications
Collect, stabilize, process, transport, and archive blood samples with Non-Indicating FTA Classic Cards. | |
Blood | |
Forensics, Transgenic identification, Transfusion medicine/HLA typing, Plasmid screening, Food and agriculture testing, Drug discovery, Genomics, STR analysis, Animal identification, Whole genome amplification, Molecular biology | |
FTA Classic Card |
Card | |
Direct STR amplification | |
4 | |
125 mL |
Safety and Handling
Signal Word
Hazard Category
- Warning
Hazard Category
- Acute toxicity Category 4