Learn More
Thermo Scientific™ Nonsterile Pipette Tips for Beckman™ Liquid Handling Systems
GREENER_CHOICE
Increase pipetting performance through the superior tip straightness and low %CV of these pipette tips for automated liquid handling systems.
Brand:  Thermo Scientific™ 918-252
650.51 EUR valid until 2024-12-31
Use promo code "21898" to get your promotional price.
 View more versions of this product
Product Code. 11711263
  • 849.00 € / Pack of 4800
Estimated Shipment: 08-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Increase pipetting performance with Thermo ScientificTM Nonsterile Pipette Tips for BeckmanTM Liquid Handling Systems, which undergo a rigorous 15-point quality control program to ensure consistency. They provide accurate and precise performance because of superior tip straightness and a low coefficient of variation. The pipette tips are validated on the actual workstations for which they are created.
Features:
  • Available in low retention and extended length
  • Lot-tested during production on actual workstation for which they are created
  • Extended length tips compatible with 96-, 384- and 1536- DeepWell™ plates
  • Low percentage coefficient of variation (%CV)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
20 μL
Non-sterile
Conductive
Beckman
Non-filtered
Rack
Beckman Coulter Biomek FX, Beckman Coulter Biomek NX SPAN-8
Certified Quality